Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1921

Sigma-Aldrich

Anti-Laminin β1 Antibody, clone 4E10

ascites fluid, clone 4E10, Chemicon®

Synonym(s):

Laminin B1 Chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

clone

4E10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Specificity

Reacts with the B1 portion of intact laminin and blocks the neurite stimulating activity of laminin.

Immunogen

Purified human laminin

Application

Anti-Laminin β1 Antibody, clone 4E10 detects level of Laminin β1 & has been published & validated for use in ELISA, IH, IP & WB.
ELISA (50% maximal binding to human laminin): >1:1,600,000.

Immunofluorescence

Affinity chromatography

Immunoprecipitation

Immunoblotting

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service