MAB898
Anti-Coxsackievirus (Echovirus 34), Clone 550-3A (1mg) KC
Synonym(s):
Anti-Coxsackievirus 24 (Echovirus 34)
About This Item
biological source
mouse
Quality Level
clone
550-3A, monoclonal
monoclonal
species reactivity
human
technique(s)
immunofluorescence: suitable
storage temp.
2-8°C
Application
Dilute with buffer pH 7.5 -8.0 containing protein or other stabilizing medium.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service