Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1722

Sigma-Aldrich

Anti-PTEN/PTEN-Long Antibody, clone 1A1

clone 1A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

clone

1A1, monoclonal

Quality Level

isotype

IgG1κ

Specificity

Clone 1A1 is a mouse monoclonal antibody that detects human PTEN. It can target both PTEN and PTEN-Long.

Immunogen

A bacterial fusion protein with a V5/His-tag at its C- terminus.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service