Skip to Content
MilliporeSigma

MABE1815

Anti-p27 Antibody, clone SON82

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

conjugate

unconjugated

antibody form

purified antibody

clone

SON82
monoclonal

purified by

using protein G

species reactivity

rat, mouse, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable

isotype

IgG2aκ

UniProt accession no.

target post-translational modification

unmodified

General description

22.19 kDa calculated.
His-tagged full-length recombinant murine p27 protein.

Application

Immunohistochemistry (Paraffin) Analysis: A 1:25 dilution from a representative lot detected p27 in mouse brain tissue sections.

Immunohistochemistry (Paraffin) Analysis: A representative lot detected p27 in Immunohistochemistry applications (Munoz-Espin, D., et. al. (2013). Cell. 155(5):1104-18).

Biochem/physiol Actions

SON82 is a rat monoclonal antibody that specifically detects mouse p27Kip1 protein.

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Immunohistochemistry (Paraffin) in mouse testis tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:25 dilution of this antibody detected p27 in mouse testis tissue sections.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service