MABF2245
Anti-PVR (CD155) Antibody, clone SKII.4
Sign Into View Organizational & Contract Pricing
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Skip To
Pricing and availability is not currently available.
conjugate
unconjugated
antibody form
purified antibody
clone
SKII.4
monoclonal
purified by
using protein G
species reactivity
human, human, mouse
concentration
(Please refer to lot specific datasheet.)
technique(s)
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
isotype
IgG1κ
UniProt accession no.
mode of action
| inhibits
target post-translational modification
unmodified
General description
SK-N-S1 human neuroblastoma cells
Specificity
Clone SKII.4 is a mouse monoclonal antibody that specifically detects human PVR (CD155).
Application
Epitope: extracellular domain
Quality
Evaluated by Flow Cytometry in HeLa cells.
Flow Cytometry Analysis: 1 µg of this antibody detected PVR (CD155) in HeLa cells.
Flow Cytometry Analysis: 1 µg of this antibody detected PVR (CD155) in HeLa cells.
Physical properties
45.31 kDa calculated.
Physical form
Purified mouse monoclonal antibody IgG1 in PBS without azide.
Storage and Stability
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service