Skip to Content
MilliporeSigma

MABF2271

Anti-BLAST-2 (CD23), clone EBVCS2

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

mol wt

(36.47 kDa calculated.)

purified by

using protein G

species reactivity

human, mouse

isotype

IgG1, kappa

Protein ID accession no.

UniProt accession no.

target post-translational modification

unmodified

Application

Anti-BLAST-2 (CD23), clone EBVCS2, Cat. No. MABF2271, is a mouse monoclonal antibody that detects Low affinity immunoglobulin epsilon Fc receptor (BLAST-2) and is tested for use in Flow Cytometry and Immunoprecipitation.
Flow Cytometry Immunoprecipitation
Immunoprecipitation Analysis: A representative lot immunoprecipitated BLAST-2 (CD23) in Immunoprecipitation applications (Sugden, B., et. al. (1983). J Virol. 46(3):800-7).

Flow Cytometry Analysis: A representative lot detected BLAST-2 (CD23) in Flow Cytometry applications (Balan, S., et. al. (2014). J Immunol. 193(4):1622-35).

Biochem/physiol Actions

Clone EBVCS2 is a mouse monoclonal antibody that detects Low affinity immunoglobulin epsilon Fc receptor (CD23; BLAST-2).

Features and Benefits

EBV-transformed human cells.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Other Notes

Evaluated by Flow Cytometry in Raji cells.

Flow Cytometry Analysis: 2 µg of this antibody detected BLAST-2 (CD23) in one million Raji cells.
Low affinity immunoglobulin epsilon Fc receptor (UniProt: P06734; also known as BLAST-2, C-type lectin domain family 4 member J, Fc-epsilon-RII, Immunoglobulin E-binding factor, Lymphocyte IgE receptor, CD23) is encoded by the FCER2 (also known as CD23A, CLEC4J, FCE2, IGEBF) gene (Gene ID: 2208) in human. Epstein-Barr virus (EBV) is reported to transform human B lymphocytes and is a causative agent for Burkitt s lymphoma and infectious mononucleosis. EBV induces the expression of one or more antigens (e.g., CD23) on the surface of adult human B-lymphocytes soon after infection. CD23 is a homotrimeric, single-pass type II membrane protein that serves as a low-affinity receptor for Low-affinity receptor for immunoglobulin E (IgE) and CR2/CD21 and plays an essential role in the regulation of IgE production and in the differentiation of B-cells. CD23 can also exist as a soluble excreted form, sCD23 (aa 150-321), which is produced by ADAM10-mediated ectoderm shedding. CD23 has a cytoplasmic domain (aa 1-21), a transmembrane domain (aa 22-47), and an extracellular domain (aa 48-321). Clone EBVCS2 can specifically detect antigens that are present on the surface of EBV-transformed cells. These antigens are also recognized by autochthonous cytotoxic T cells specific for EBV-transformed cells. (Ref.: Kintner, C., and Sugden, B. (1981). Nature. 294(3); 458-460).
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service