Skip to Content
MilliporeSigma

MABF2308

Anti-HCV E2 Antibody, clone 19B3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
19B3, monoclonal
Application:
ELISA, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

unconjugated

antibody form

purified antibody

clone

19B3, monoclonal

purified by

using protein G

species reactivity

mouse, virus

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable, western blot: suitable

isotype

IgG2aκ

capacity

neutralization

UniProt accession no.

target post-translational modification

unmodified

General description

327.15 kDa calculated for the entire genome polyprotein.
DNA constructs of E1E2 followed by a boost with truncated E2 protein (aa 412-645).

Application

ELISA Analysis: A representative lot detected HCV E2 in ELISA applications (Ruwona, T.B., et. al. (2014). J Virol. 88(18):10459-71; Aleman, F., et. al. (2018). Proc Natl Acad Sci USA. 115(29):7569-7574).
Western Blotting Analysis: A representative lot detected HCV E2 in Western Blotting applications (Chen, F., et. al. (2019). Methods Mol Biol. 1911:295-304).

Neutralizing Activity: A representative lot displayed high neutralizing activity against HCV isolates. (Aleman, F., et. al. (2018). Proc. Natl. Acad. Sci. USA. 115(29); 7569-7574; Ruwona, T.B., et. al. (2014). J. Virol. 88(18); 10459-71).


ELISA Analysis: A representative lot detected HCV E2 in ELISA applications (Ruwona, T.B., et. al. (2014). J Virol. 88(18):10459-71; Aleman, F., et. al. (2018). Proc Natl Acad Sci USA. 115(29):7569-7574).

Biochem/physiol Actions

Clone 19B3 is a mouse monoclonal antibody that detects the envelope glycoprotein E2 of the Hepatitis C virus. It targets an epitope within 12 amino acids from the N-terminal region of the E2.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without preservatives.

Preparation Note

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by ELISA in H77 E1E2-transfected 293T cell lysate.

ELISA Analysis: This antibody detected HCV E2 in H77 E1E2 -transfected 293T cell lysate, but not in control cell lysate.

Other Notes

Virus (Hepatitis C virus).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library