Sign Into View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Skip To
conjugate
unconjugated
antibody form
purified antibody
clone
N416/40
monoclonal
purified by
using protein G
species reactivity
mouse, rat, mouse
concentration
(Please refer to lot specific datasheet.)
technique(s)
immunohistochemistry: suitable
isotype
IgG2aκ
UniProt accession no.
target post-translational modification
unmodified
General description
127.54 kDa calculated.
Linear peptide from the N-terminal region of mouse Glutamate receptor ionotropic, NMDA3A (NR3A).
Application
Epitope: extracellular domain
Biochem/physiol Actions
Clone N416/40 is a mouse monoclonal antibody that detects NMDAR3A (NR3A).
Physical form
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Immunohistochemistry (Paraffin) in rat brain tissue sections.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected NR3A in rat brain tissue sections.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected NR3A in rat brain tissue sections.
Other Notes
Mouse, Rat
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service