Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN2307

Sigma-Aldrich

Anti-TTR mutant (Y78F) Antibody, clone AD7

new

clone AD7 (AD7F6)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

clone

AD7 (AD7F6), monoclonal

Quality Level

isotype

IgG2b

Specificity

Clone AD7 is a mouse monoclonal antibody that specifically detects glycosylated form of TTR with Y78F mutation. Does not react with TTR bearing F33V, V30A, and K70N mutations.

Immunogen

Recombinant human Transthyretin with Y78F mutation.

Reconstitution

1 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service