MABS1949
Anti-NG2 Antibody, clone 1E6
About This Item
Skip To
conjugate
unconjugated
antibody form
purified antibody
clone
1E6
monoclonal
purified by
using protein G
species reactivity
rat, mouse, mouse
concentration
(Please refer to lot specific datasheet.)
technique(s)
ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG1κ
UniProt accession no.
target post-translational modification
unmodified
General description
Specificity
Application
Western Blotting Analysis: A representative lot detected NG2 in Western Blotting applications (Stegmuller, J., et. al. (2002). J Neurocytol. 31(6-7):497-505; Schneider, S., et. al. (2001). J Neurosci. 21(3):920-33; Niehaus, A., et. al. (1999). J Neurosci. 19(12):4948-61; Trotter, J., et. al. (2005). J Neurol Sci. 233(1-2):37-42; Chatterjee, N., et. al. (2008). J Biol Chem. 283(13):8310-7).
ELISA Analysis: A representative lot detected NG2 in ELISA applications (Maus, F., et. al. (2015). PLoS One. 10(9):e0137311).
Immunofluorescence Analysis: A representative lot detected NG2 in Immunoprecipitation applications (Schneider, S., et. al. (2001). J Neurosci. 21(3):920-33; Niehaus, A., et. al. (1999). J Neurosci. 19(12):4948-61; Karram, K., et. al. (2005). J Anat. 207(6):735-44; Chatterjee, N., et. al. (2008). J Biol Chem. 283(13):8310-7).
Immunohistochemistry Analysis: A representative lot detected NG2 in Immunohistochemistry applications (Karram, K., et. al. (2005). J Anat. 207(6):735-44; Niehaus, A., et. al. (1999). J Neurosci. 19(12):4948-61).
Quality
Western Blotting Analysis: 2 µg/mL of this antibody detected NG2 in mouse brain tissue lysates.
Physical properties
Physical form
Storage and Stability
Disclaimer
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service