Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2185

Sigma-Aldrich

Anti-NPHP1 Antibody, clone 1M4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

conjugate

unconjugated

antibody form

purified antibody

clone

1M4
monoclonal

purified by

using protein G

species reactivity

human, human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG3κ

UniProt accession no.

target post-translational modification

unmodified

General description

A synthetic polypeptide fused by 16 different NPHP1 peptide sequences predicted to have the highest antigenicity. The sequence was converted to cDNA and the polypeptide was expressed and purified from E. coli.

Specificity

Clone 1M4 is a mouse monoclonal antibody that specifically detects human Nephrocystin-1 (NPHP1).

Application

Immunofluorescence Analysis: A representative lot of this antibody detected NPHP1 in Immunofluorescence applications (Xu, Q., et. al. (2016). Nat Commun. 7:10777).

Quality

Evaluated by Western Blotting in human testis tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected NPHP1 in human testis tissue lysate.

Physical properties

~92 kDa observed; 83.30 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified mouse monoclonal antibody IgG3 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service