Skip to Content
MilliporeSigma

MABX4130-10KC

Anti-human PAPP-A (dPAPP-A) clone 30

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
30, monoclonal
Species reactivity:
mouse, human
Application:
ELISA, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody form

purified antibody

Quality Segment

clone

30, monoclonal

purified by

using Protein A

species reactivity

mouse, human

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

ELISA: suitable, western blot: suitable

isotype

IgG1

General description

dPAPP-A

Application

Confirmed matched pairing. Capture Antibody MABX4152-1KC; Detector Antibody MABX4130-1KC

Physical form

PBS (pH 7.4) with 0.09% sodium azide

Preparation Note

Stable for 12 months at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library