Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX4152-10KC

Sigma-Aldrich

Anti-human PAPP-A (dPAPP-A) clone 52

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$11,120.00

$11,120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$11,120.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$11,120.00


Check Cart for Availability

Request a Bulk Order

antibody form

purified antibody

Quality Level

clone

52
monoclonal

purified by

using Protein A

species reactivity

human, mouse

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

General description

dPAPP-A

Application

Confirmed matched pairing. Capture Antibody MABX4152-1KC; Detector Antibody MABX4130-1KC

Physical form

PBS (pH 7.4) with 0.09% sodium azide

Storage and Stability

Stable for 12 months at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service