MABX8281
Anti-Hepatitis B e Antigen (HBeAg) Antibody, (1mg)
Sign Into View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
Skip To
antibody form
purified antibody
clone
1
monoclonal
purified by
ion-exchange chromatography
species reactivity
mouse, virus
concentration
(Please refer to lot specific datasheet.)
technique(s)
ELISA: suitable
immunodiffusion: suitable
isotype
IgG1
General description
Native HBeAg
Biochem/physiol Actions
e antigen to Hepatitis B virus
Physical form
Purified mouse monoclonal antibody in buffer containing 10 mM Tris-HCl, pH 8.0, 0.05% NaN3
Preparation Note
2-8C. For long-term storage, freeze at -20C.
Analysis Note
Detect HBeAg using mouse monoclonal Anti-HBeAg, Cat No MABX8281-1KC, validated for use in ELISA
Other Notes
e antigen of Hepatitis B virus
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service