Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 ea |
About This Item
This item is not able to be purchased as a guest user. For further information, please contact us at +1 866 227 3811or
.Product Name
Millistak+® Pod Depth Filter, CE30 (media series), cellulose membrane, effective filtration area 1.4 m2
Quality Segment
material
TPE (Thermo Plastic Elastomer), TPE gasket (Thermo Plastic Elastomer), cellulose membrane, polypropylene housing (glass-filled)
sterilization compatibility
autoclavable compatible
effective filtration area
1.4 m2
fitting
seal (flat seal)
cartridge code
CE30 (media series)
General description
This CE30 media series delivers optimal performance through a gradient density matri, ideal for primary (coarse) clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
Features and Benefits
- Low hold-up volume for greater product yield
- Broad range of media types offered in single and multilayer products
- Flexible, modular format offers scalability from 5 to 12,000 liters or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Self-contained Pod filters protect operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| fitting seal (flat seal) | fitting seal (flat seal) | fitting seal (flat seal) | fitting seal (flat seal) |
| material TPE (Thermo Plastic Elastomer), cellulose membrane, TPE gasket (Thermo Plastic Elastomer), polypropylene housing (glass-filled) | material TPE (Thermo Plastic Elastomer), TPE gasket (Thermo Plastic Elastomer), cellulose membrane, polypropylene housing (glass-filled) | material TPE (Thermo Plastic Elastomer), TPE gasket (Thermo Plastic Elastomer), cellulose membrane, polypropylene housing (glass-filled) | material TPE (Thermo Plastic Elastomer), TPE gasket (Thermo Plastic Elastomer), cellulose membrane, polypropylene housing (glass-filled) |
| cartridge code CE30 (media series) | cartridge code CE20 (media series) | cartridge code CE20 (media series) | cartridge code CE45 (media series) |
| effective filtration area 1.4 m2 | effective filtration area 1.4 m2 | effective filtration area 0.77 m2 | effective filtration area 1.4 m2 |
| sterilization compatibility autoclavable compatible | sterilization compatibility autoclavable compatible | sterilization compatibility autoclavable compatible | sterilization compatibility autoclavable compatible |
| Quality Level 400 | Quality Level 400 | Quality Level 400 | Quality Level 400 |
Still not finding the right product?
Explore all of our products under Millistak+® Pod Depth Filter
Storage Class
10-13 - German Storage Class 10 to 13
Stay compliant with GMP using the Emprove® Program
Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.
Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.
Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite
Not finding what you are looking for? Visit the Emprove® Program Help page.