MCE4501FS1
Millistak+® Pod Depth Filter
CE45, filtration area 0.11m², Process Scale
Select a Size
$323.00
Select a Size
About This Item
Skip To
$323.00
Product Name
Millistak+® Pod Depth Filter, CE45 (media series), cellulose membrane, effective filtration area 0.11 m2
material
TPE (Thermo Plastic Elastomer)
TPE gasket (Thermo Plastic Elastomer)
cellulose membrane
polypropylene housing (glass-filled)
Quality Level
sterilization compatibility
autoclavable compatible
effective filtration area
0.11 m2
fitting
seal (flat seal)
cartridge code
CE45 (media series)
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | MCE4513FS1 | MCE4013FS1 | MCE3013FS1 |
---|---|---|---|
fitting seal (flat seal) | fitting seal (flat seal) | fitting seal (flat seal) | fitting seal (flat seal) |
material TPE (Thermo Plastic Elastomer), cellulose membrane, TPE gasket (Thermo Plastic Elastomer), polypropylene housing (glass-filled) | material TPE (Thermo Plastic Elastomer), TPE gasket (Thermo Plastic Elastomer), cellulose membrane, polypropylene housing (glass-filled) | material TPE (Thermo Plastic Elastomer), TPE gasket (Thermo Plastic Elastomer), cellulose membrane, polypropylene housing (glass-filled) | material TPE (Thermo Plastic Elastomer), TPE gasket (Thermo Plastic Elastomer), cellulose membrane, polypropylene housing (glass-filled) |
sterilization compatibility autoclavable compatible | sterilization compatibility autoclavable compatible | sterilization compatibility autoclavable compatible | sterilization compatibility autoclavable compatible |
effective filtration area 0.11 m2 | effective filtration area 1.4 m2 | effective filtration area 1.4 m2 | effective filtration area 1.4 m2 |
cartridge code CE45 (media series) | cartridge code CE45 (media series) | cartridge code CE40 (media series) | cartridge code CE30 (media series) |
Quality Level 400 | Quality Level 400 | Quality Level 400 | Quality Level 400 |
General description
This CE45 media series delivers optimal performance through a gradient density matri, ideal for primary (coarse) clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
Features and Benefits
- Low hold-up volume for greater product yield
- Broad range of media types offered in single and multilayer products
- Flexible, modular format offers scalability from 5 to 12,000 liters or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Self-contained Pod filters protect operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Legal Information
Storage Class
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service