MilliporeSigma
All Photos(1)

Documents

ME-BOOST-TR

Millipore

MicroEssentials Boost-TR

packaging

carton of 10 kg

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

General description

MicroEssentials Boost-TR is a time-release yeast nutrient specially designed for wine fermentations. The product consists of 1.5 inch tablets intended for direct addition to juices or musts. The unique time-release system provides high levels of inorganic nitrogen and low levels of organic nitrogen, vitamins and trace minerals in a sustained manner, thus assuring timely supplementation and proper management of the fermentation.

The convenience of MicroEssentials Boost-TR offers winemakers ultimate control in supplementing juices and musts throughout fermentation with one single addition. Utilizing MicroEssentials Boost-TR offers similar fermentation kinetics compared with incremental nutrient additions.

Application

MicroEssentials Boost-TR is suggested as a supplement to MicroEssentials Complete-TR when additional inorganic nitrogen is desired. MicroEssentials Boost-TR can be used separately in easy fermentation scenarios when mainly inorganic nitrogen supplementation is required. MicroEssentials Boost-TR is recommended for use in both red and white juices and musts. This product was designed for ease of use; therefore, to achieve full nutritional benefits, MicroEssentials Boost-TR should be added at yeast inoculation. MicroEssentials Time-Release Technology eliminates the need for multiple additions.

This product is ideal for winemakers who traditionally supplement with diammonium phosphate. MicroEssentials Boost-TR provides 19ppm available nitrogen at 1 lb / 1,000 gal.

Packaging

10 kg carton
Dose Rate: 12 – 96 g/hl (1 – 8 lbs / 1,000 gal)

Other Notes

Supplement to MicroEssentials Complete-TR when additional inorganic nitrogen is desired.

Legal Information

MicroEssentials Boost-TR is a trademark of Gusmer Enterprises, Inc
MicroEssentials Complete-TR is a trademark of Gusmer Enterprises, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service