MRSMCTS48

Millipore

Milliflex Cassete w/Tryptic Soy Agar for Milliflex Rapid

Medium used on Milliflex Rapid for the recovery of a broad range of heterotrophic microorganisms such as common aerobic and facultative anaerobic bacteria found in various types of water sources.

eCl@ss:
32160801
NACRES:
NA.74

Quality Level

form

plate (pre-poured agar)

packaging

cassette of 10 mL
pack of 48 tests

manufacturer/tradename

Milliflex®

application(s)

bioburden testing: suitable
microbiology: suitable
water monitoring: suitable

color

light amberslightly opalescent

input

sample type bottled water
sample type cosmetics
sample type pharmaceutical(s)
sample type beer

Featured Industry

Cosmetics
Pharmaceutical

compatibility

for use with Milliflex®

shipped in

wet ice

Application

Type of Test
Bioburden

Microbial Limits

Specifications

Incubation Temperature: 30–35 °C

Analysis Note

Clear to white colonies, some may produce pigment

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

WGK Germany

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.