Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MSP010060

Millipore

Funnel Adapter

for use with Milliflex®, for use with EZ-Fit®

Synonym(s):

Manifold Adapter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

material

stainless steel adapter

Quality Level

application(s)

food and beverages
pharmaceutical

compatibility

for use with EZ-Fit®
for use with Milliflex®

General description

Funnel Adapter is designed to fit Milliflex® and EZ-Fit® filtration funnels to manifolds. Microbial Filtration is a regular lab practice to carry out bioburden and water testing.
The Funnel Adapter will fit the filtration manifold with perfection and ease. Please contact our sales team for a Demo Request.

Application

Funnel Adapter is used to fit filtration funnels to manifolds during microbial filtration. Microbial filtration is an essential step to carry out bioburden and water testing.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service