Skip to Content
MilliporeSigma

MX0SP06CL9

Millistak+® HC Pro Depth Filter in NanoPod NP6 format

new

X0SP, filtration area 6 cm², Process Development Scale

Synonym(s):

6 cm2 surface area, Screening device

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

filter aid (Synthetic filter media)
polypropylene housing

Quality Level

product line

Millistak+®

feature

hydrophilic

diam.

3.6 cm (1.4 in.)

filtration area

6 cm2

impurities

<0.25 EU/mL (LAL test, clot test technique (on filter media only))

input

sample type liquid

fitting

Luer fitting

application(s)

pharma/biopharma processes

filter grade

X0SP

shipped in

ambient

General description

The Nanopod NP6 with its 6cm2 surface area is the small scale screening filter of choice for benchtop virus prefiltration and secondary clarification applications.

Application

Process Development.

Features and Benefits

Nanopod NP6 devices offer:
  • Very small surface area for low feedstream volume requirements.
  • Enables process development of virus clearance prefiltration with minimal feedstock requirements.

Packaging

9/pack - FSC certified packaging.

Other Notes

Integrity is maintained after 2 cycles of 60 minutes at 123 °C, however filtration performance may be impacted post autoclave. Recommended for post-use decontamination only.

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service