MilliporeSigma
All Photos(1)

MXRPDAG09

Millipore

Milliflex Rapid Cleaning/decontamination kit

manufacturer/tradename

Milliflex®

Quality Level

input

sample type pharmaceutical(s)
sample type beverage(s)
sample type food(s)

application(s)

bioburden testing
clinical
environmental
food and beverages
industrial qc
pharmaceutical
sterility testing
water monitoring

compatibility

Milliflex® Rapid

shipped in

ambient

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
MXRPBLRSTMXRP2SPRKTMXRPC0VER
compatibility

Milliflex® Rapid

compatibility

Milliflex® Rapid

compatibility

for use with Milliflex® Rapid

compatibility

for use with Milliflex® Autospray System

shipped in

ambient

shipped in

wet ice

shipped in

ambient

shipped in

-

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

input

sample type pharmaceutical(s)
sample type beverage(s)
sample type food(s)

input

sample type pharmaceutical(s)

input

sample type pharmaceutical(s)

input

-

Quality Level

100

Quality Level

300

Quality Level

-

Quality Level

-

General description

The AutoSpray Station issues a warning message across the control panel when calibration, cleaning or priming are required ensuring consistent and reliable performance throughout the life of the system. A self-cleaning unit, the AutoSpray Station is ready for re-use in five minutes. The unit should be recalibrated after every 5000 samples, or at least once a year.

Application

General microbiological analysis

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service