MilliporeSigma
All Photos(1)

Documents

MXRPEZFIT

Millipore

Milliflex® Autospray Station Silicone Tubes

For Liquid Reagent Transfer to Glass Nebulizer

Synonym(s):
Silicone Tubes

material

polyurethane

packaging

pkg of 2 units

manufacturer/tradename

Milliflex®

parameter

15-40 °C temp. limit

application(s)

industrial qc
microbiology
pharmaceutical
sterility testing

compatibility

for use with Milliflex® Autospray System

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
MXRPGLASSMXRPAIRTUMXRP2SPRKT
Silicone air tubes Spare part for Milliflex® Rapid AutoSpray Station

Millipore

MXRPAIRTU

Silicone air tubes

packaging

pkg of 2 units

packaging

pkg of 1 unit

packaging

pkg of 2 units

packaging

pkg of 1 kit

application(s)

industrial qc
microbiology
pharmaceutical
sterility testing

application(s)

industrial qc
microbiology
pharmaceutical
sterility testing

application(s)

food and beverages
industrial qc
pharmaceutical

application(s)

bioburden testing
food and beverages
pharmaceutical
sterility testing
water monitoring

compatibility

for use with Milliflex® Autospray System

compatibility

for use with Milliflex® Autospray System

compatibility

AutoSpray Station

compatibility

for use with Milliflex® Rapid

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

parameter

15-40 °C temp. limit

parameter

15-40 °C temp. limit

parameter

-

parameter

15-40 °C temperature

General description

Milliflex® Autospray Station Silicone Tubes are connected between vials supports and glass nebulizers inlet and designed to direct the liquid reagents from the vials to the calibrated glass nebulizer. The glass nebulizer sprays the reagent onto the filter holder containing the filtered sample. In auto-spray mode the reagents are sprayed onto the sample in 90 seconds.
Milliflex® Autospray Station is used for rapid microbial detection in filterable samples.

Application

Milliflex® Autospray Station Silicone Tubes is used to direct the Milliflex® Autospray reagents to the calibrated glass nebulizer for rapid microbial detection.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service