Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MXRPGLASS

Millipore

Milliflex® Autospray Station Glass Nebulizer

For Spraying Reagents on the Milliflex® membrane

Synonym(s):

Calibrated Glass Nebulizer, Glass Nebulizer

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$3,360.00

$3,360.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 UNIT
$3,360.00

About This Item

UNSPSC Code:
41116500
NACRES:
NB.79

$3,360.00


Check Cart for Availability

Request more information

material

polyurethane

packaging

pkg of 1 unit

manufacturer/tradename

Milliflex®

parameter

15-40 °C temp. limit

application(s)

industrial qc
microbiology
pharmaceutical
sterility testing

compatibility

for use with Milliflex® Autospray System

General description

Milliflex® Autospray Station Glass Nebulizer is an integral part of the Milliflex® Rapid Detection System which is calibrated for its use. The glass nebulizer part is responsible to spray the rapid detection reagents onto the sample though an automated program. The internal time-control pinch valve ensures the precise volume of reagent being dispensed through calibrated glass nebulizer at every dispensing cycle.
Milliflex® Autospray Station is used for rapid microbial detection in filterable samples.

Application

Milliflex® Autospray Station Glass Nebulizer is used to spray pre-set volumes of rapid microbial detection reagents on filtered samples. The calibrated glass nebulizer dispenses exact volumes at each dispense cycle.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service