MXRPSPFH1
Filter Holder
for use with Milliflex® Rapid Autospray station
Synonym(s):
MXRP SPF H1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.79
material
316 stainless steel holder
product line
Milliflex®
packaging
units of 1
manufacturer/tradename
Milliflex®
compatibility
for use with Milliflex® Rapid Autospray station
General description
Filter Holder for Milliflex® Rapid Autospray station is an accessory which is designed to evenly spray the Milliflex® Rapid reagent onto the membrane filter before rapid detection by the Milliflex® Rapid Detection System.
The Milliflex Rapid Microbiology Detection and Enumeration system is an automated solution for the rapid detection, response, and resolution of microbial contamination in filterable samples throughout the manufacturing process. The system improves process control, product yield and the timely release of products. Based on Adenosine Triphosphate (ATP) bioluminescence technology, the Milliflex Rapid system delivers faster test results than traditional microbial contamination detection methods, such as membrane filtration (MF) and pour plates. The Milliflex Sample Prep method also ensures consistent, reliable results. The Millilfex Rapid system can clearly distinguish between mixed microbial growth of slow growing and fast growing microorganisms, as well as variances in their size and ATP content in water samples.
Application
Rapid Testing;Pharmaceutical Products Analysis
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service