MilliporeSigma
  • NE1037
All Photos(1)

NE1037

Sigma-Aldrich

Anti-APOC1 Mouse mAb (2E2-1A3)

liquid, clone 2E2-1A3, Calbiochem®

Synonym(s):
Anti-Apolipoprotein

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E2-1A3, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... APOC1(341)

General description

Anti-APOC, mouse monoclonal, clone 2E2-1A3, recognizes the ~10 kDa APOC1 protein in human liver tissue. It is validated for ELISA and Western blotting.
Purified mouse monoclonal antibody. Recognizes the ~10 kDa APOC1 protein.
Recognizes the ~10 kDa APOC1 protein in human liver tissue.

Immunogen

A recombinant polypeptide corresponding to amino acids 1-84 of human APOC1, expressed as a GST fusion protein

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Andersson, B., et al. 2009. Proteome Sci.7, 40.
Meunier, J.C., et al. 2008. J. Virol.82 9647.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Please enter the Product Number and Lot Number.

Certificate of Quality

Please enter the Product Number and Lot Number.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service