Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OP24

Sigma-Aldrich

Anti-c-K-Ras (Ab-1) Mouse mAb (234-4.2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

biological source

mouse

Quality Level

form

liquid

species reactivity

(human, mouse, rat)

storage condition

do not freeze

dilution

(Frozen Sections (10 µg/mL)
Immunoblotting (5 µg/mL with 1% gelatin)
Immunoprecipitation
Paraffin Sections (10 µg/mL, saponin or pepsin pre-treatment required))

isotype

IgG2a

storage temp.

2-8°C

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with Ag8.653 mouse myeloma cells. Recognizes the ~21 kDa c-K-Ras and v-K-Ras proteins. Also recognizes c-H-Ras and c-N-Ras.

Application

Frozen Sections (10 µg/mL)Immunoblotting (5 µg/mL with 1% gelatin)Immunoprecipitation Paraffin Sections (10 µg/mL, saponin or pepsin pre-treatment required)

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service