Skip to Content
MilliporeSigma

P2PLACC25

Pellicon® 2 Maxi Cassette Media: 1 kDa

Chemistry: Regenerated Cellulose (RC) Screen Type: C Device Size: 2.5 m2

Synonym(s):

Pellicon® 2 Maxi Ultrafiltration Module PLAC C 2.5 m2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32031623
UNSPSC Code:
23151806
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Pellicon® 2 Maxi Cassette Media: 1 kDa, Chemistry: Regenerated Cellulose (RC) Screen Type: C Device Size: 2.5 m2

Quality Segment

product line

Pellicon®

technique(s)

analytical sample preparation: suitable, protein purification: suitable

H

7.6 cm (3 in.)

L

21 cm (8.3 in.)

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
P2PLACC2504053252259739