Skip to Content
MilliporeSigma

Skip To

P2PLBCV05

Pellicon® 2 Cassette Media: 3 kDa

Chemistry: Regenerated Cellulose (RC) Screen Type: V Device Size: 0.5 m2

Synonym(s):

Pellicon® 2 Cassette Ultrafiltration Module PLBC V 0.5 m2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

eCl@ss:
32031623
UNSPSC Code:
23151806
This product is sold exclusively by Millipore via offline channels. Contact Customer Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Pellicon® 2 Cassette Media: 3 kDa, Chemistry: Regenerated Cellulose (RC) Screen Type: V Device Size: 0.5 m2

Quality Level

product line

Pellicon®

technique(s)

analytical sample preparation: suitable, protein purification: suitable

H

2.16 cm (0.85 in.)

L

21 cm (8.3 in.)

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P2PLBCC05P2PLACC05P2PLBCC25
product line

Pellicon®

product line

Pellicon®

product line

Pellicon®

product line

Pellicon®

technique(s)

analytical sample preparation: suitable, protein purification: suitable

technique(s)

analytical sample preparation: suitable, protein purification: suitable

technique(s)

analytical sample preparation: suitable, protein purification: suitable

technique(s)

analytical sample preparation: suitable, protein purification: suitable

Quality Level

400

Quality Level

400

Quality Level

400

Quality Level

400

L

21 cm (8.3 in.)

L

21 cm (8.3 in.)

L

21 cm (8.3 in.)

L

21 cm (8.3 in.)

H

2.16 cm (0.85 in.)

H

1.5 cm (0.6 in.)

H

1.5 cm (0.6 in.)

H

7.6 cm (3 in.)


Still not finding the right product?

Explore all of our products under Pellicon® 2 Cassette Media: 3 kDa


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
P2PLBCV0504053252313523

Questions

Reviews

No rating value

Active Filters