Skip to Content
MilliporeSigma

PBGC15005

Biomax® 10 kDa Ultrafiltration Discs

Chemistry: Polyethersulfone (PES) Filter Diameter: 150 mm Amicon® Bioseparations

Synonym(s):

Biomax 10 kDa Ultrafiltration Discs

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
40161507
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

B10K 150MM HIGH FLUX 5PK, Chemistry: Polyethersulfone (PES) Filter Diameter: 150 mm Amicon® Bioseparations

Quality Segment

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under B10K 150MM HIGH FLUX 5PK


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
PBGC1500504053252826641