PF133
TACE, His Tag, Human, Recombinant, T. ni.
Synonym(s):
ADAM17, Human α-Secretase, Apo2L; TNF-Related Apoptosis-Inducing Ligand
About This Item
Recommended Products
form
lyophilized
Quality Level
storage condition
OK to freeze
solubility
soluble (Reconstitute with 100 µl dH2O.)
storage temp.
−20°C
General description
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service