Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RX0175

Sigma-Aldrich

Rosolic Acid

Rosolic Acid

Synonym(s):

Rosolic Acid, Paraosolic Acid, Aurin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H4OH)2C=(C6H4=O)
CAS Number:
Colour Index Number:
43800
MDL number:
UNSPSC Code:
41116121
EC Index Number:
210-041-8

form

powder

loss

≤5.0% loss on drying

color

red

mp

292 °C

UV absorption

λ: 480-485 nm Amax

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

room temp

InChI

1S/C19H14O3/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15/h1-12,20-21H

InChI key

FYEHYMARPSSOBO-UHFFFAOYSA-N

Analysis Note

Color: Red
Extinction coefficient: 45000 min
Form: Powder
Loss on drying: 5.0% max
Wavelength of max. absorbance: 480-485nm
pH of a 1% aqueous solution:

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service