Skip to Content
MilliporeSigma

SAU0000L2PLP5

Millipore

Mobius®, Silver, Tubing Assy

Tubing assembly with C-Flex® 374 and Pharma APT

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

C-Flex® tubing (Precut C-Flex 374 (1/2"ID X 3/4"OD) 36"LG)
C-Flex® tubing (Precut C-Flex 374 (3/8"ID X 5/8"OD) 36"LG)
tubing (Pharma APT (1/2"ID X 3/4"OD))

Quality Level

agency

meets requirements for ISO 10993-5
meets requirements for ISO 11137
meets requirements for USP <788>
meets requirements for USP <88>

sterility

sterile; γ-irradiated (level of 25-40 kGY)

product line

Mobius®

packaging

package of 5 ×

technique(s)

buffer exchange: suitable
media fill: suitable

application(s)

cell therapy
gene therapy
life science and biopharma
liquid processing
mAb
ophthalmics
parenterals
pharma/biopharma processes
viral therapy
waste management

storage temp.

2-30°C

General description

Tubing assembly with C-Flex® 374 and PHARMA APT is the perfect solution for your operational efficiency! Designed for seamless integration into your processes, these high-quality assemblies are ready to go, saving you time and resources. With our reliable and durable materials, you can trust that your operations will run smoothly. Don′t wait—streamline your workflow today with our convenient, ready-to-use assemblies and experience the difference in performance and reliability!

Application

Liquid Processing Applications

Features and Benefits

Standard products eliminate design and manufacturing lead times.

Easily connects to other single-use assemblies.

Legal Information

C-Flex is a registered trademark of Saint-Gobain Performance Plastics Corp.
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service