Documents

SCC103

Sigma-Aldrich

HAPI Rat Microglial Cell Line

Synonym(s):
HAPI (Highly Aggressively Proliferating Immortalized) cell line
eCl@ss:
32011203

Quality Level

biological source

rat

application(s)

cell analysis: suitable (cell based assays)
cell culture | mammalian: suitable

shipped in

ambient

General description

Microglia are the primary immune cells of the central nervous system (CNS), and accounts for 10-15% of all cells found within the brain. Microglial play an important role in maintaining the health of the CNS by the removal of pathogens, infectious agents and damaged cells through phagocytosis. Microglial have also been extensively studied for their harmful roles in a variety of neurodegenerative disease including Alzheimer’s disease, Parkinson’s disease and amyotrophic lateral sclerosis.

The HAPI Rat Microglial Cell Line is a spontaneously immortalized cell line derived from primary microglia-enriched cultures prepared from 3-day-old rat brains (1). HAPI (highly aggressively proliferating immortalized) cells retain characteristics of microglia including marker expression, proinflammatory cytokine secretion in response to LPS, and phagocytic activity (1). These cells provide an alternative model to primary microglial cells.

Reference:
1. Cheepsunthorn, P., Radov, L., Menzies, S., Reid, J., & Connor, J. R. (2001). Characterization of a novel brain‐derived microglial cell line isolated from neonatal rat brain. Glia, 35(1), 53-62.

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells tested negative for infectious diseases by a Mouse/Rat Comprehensive CLEAR panel PCR panel by Charles River Animal Diagnostic Services.
• Cells tested negative for mycoplasma contamination.
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.