Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
23151806
Quality Segment
Storage Class
4.1B - Flammable solid hazardous materials
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
PrefilterFamilyGuide
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SH1J033H03 | 04053252881824 |