Skip to Content
MilliporeSigma

SK1P006W6

PEL 2 B30 C SCREEN 5 SQFT

Synonym(s):

PEL 2 B30 C SCREEN 5 SQFT

Sign In to View Organizational & Contract Pricing.

Select a Size

This product is sold exclusively by Millipore via offline channels. Contact Customer Service

About This Item

UNSPSC Code:
23151806

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Pellicon® 2 Cassette Media: BIOMAX® 30 kDa, Chemistry: Polyethersulfone (PES) Screen Type: C Device Size: 0.5 m²

Quality Level

quality

400

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service