Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SK2P218W2

Millipore

CDUF050LB PLBC SS50 1EA

Sign Into View Organizational & Contract Pricing


About This Item

This product is sold exclusively by Millipore via offline channels. Contact Customer Service

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service