Skip to Content
MilliporeSigma

SN4M002A04

Millipore

Mobius®, Silver, 500ML PF Bag Assy, Lynx® ST

500ML Pureflex® Bag Assembly with Lynx® ST Connection

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

PureFlex device (film)
connections (Lynx ST)

Quality Level

sterility

γ-irradiated (level of 25-40 kGY)

product line

Mobius®

packaging

package of 1 ×

technique(s)

buffer exchange: suitable
media fill: suitable

fitting

connector (Lynx ST 1/4"HB X 3/4"TC (STC11FHA10))
female Luer connector (Luer Female SmartSite Needleless Valve)
3/16 in. female Luer connector (Luer Female 3/16" HB PVDF)
plug (plug-end 3/16"HB)

application(s)

cell therapy
gene therapy
life science and biopharma
liquid processing
mAb
ophthalmics
parenterals
pharma/biopharma processes
viral therapy
waste management

storage temp.

2-30°C

General description

500ML Pureflex Bag Assembly with Lynx® ST Connection

Application

Liquid Processing Applications

Features and Benefits

Standard products eliminate design and manufacturing lead times.Utilizes PureFlex Plus Film and components that can easily scale up.

Legal Information

LYNX is a registered trademark of Merck KGaA, Darmstadt, Germany
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
PUREFLEX is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service