MilliporeSigma
SPKIPBINS028
Spare parts for Mobius® iFlex Bioreactor
SU DO1&DO2 Holder
Sign In to View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
41104909
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
stainless steel
Quality Level
product line
Mobius®
packaging
pkg of 1 unit
compatibility
for use with Millipore Mobius®
General description
The Mobius® iFlex Bioreactor is a next-generation, single-use, modular bioreactor meticulously engineered for optimal performance in both batch and perfusion processes.
Application
Spare parts for Mobius® iFlex Bioreactor
Features and Benefits
Seamless Transition from Batch to Perfusion Processes
Other Notes
Our recommendation is to perform a risk-based review of all spare parts documented in the Mobius(r) iFlex system′sspare parts list, based on equipment uptime requirements. Parts assessed as critical for the functioningof your system should be kept in your stock to have them readily available in case they need to bereplaced.
Legal Information
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Buying original Mobius® IFlex spare parts to maintain optimal performance and minimize downtime