Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ST1598

Sigma-Aldrich

Anti-YWHAG Mouse mAb (6A10)

liquid, clone 6A10, Calbiochem®

Synonym(s):

Anti-Tyrosine 3-Monooxygenase/Tryptophan5-Monooxygenase Activation Protein γ polypeptide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6A10, monoclonal

form

liquid

does not contain

preservative

species reactivity

rat, human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Ywhag(22628)

General description

Purified mouse monoclonal antibody. Recognizes the ~25 kDa YWHAG protein.
Recognizes the ~25 kDa YWHAG protein in PC-12, IMR32, NIH3T3, and RAW 264.7 cells.
This Anti-YWHAG Mouse mAb (6A10) is validated for use in ELISA, Immunoblotting for the detection of YWHAG.

Immunogen

A recombinant polypeptide corresponding to amino acids 70-170 of human YWHAG, expressed as a GST fusion protein

Warning

Toxicity: Regulatory Review (Z)

Analysis Note

Positive Control
PC-12 cells, IMR32 cells, NIH3T3 cells, RAW 264.7 cells

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service