MilliporeSigma
All Photos(4)

STBMRFA1

Millipore

Fluid A

pkg of 100 mL (Bottle with screw cap and septum), For rinsing and dilution, suitable for sterility testing, for use with Steritest®

Synonym(s):
Rinse Fluid, Washing Fluid
eCl@ss:
32014001
NACRES:
NA.74

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

ready-to-use

packaging

pkg of 100 mL (Bottle with screw cap and septum)

bottle volume

100 mL , Filling Volume

color

clear, with no precipitate and free of visible particles

input

sample type pharmaceutical(s)

pH

7.1±0.2 (25.0 °C)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest®

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Fluid A is a general rinse buffer in compliance with the European Pharmacopeia (EP) 2.6.1, United States Pharmacopeia (USP) 71 and Japanese Pharmacopeia (JP) 4.06. The product is a clear colorless liquid with a pH of 7.1 ± 0.2 at 25°C. It can be stored at 2-25°C (protected from light) for 12 months. Each bottle has a screw cap with septum closure to reduce the risk of cross-contamination and optimize the disinfection procedures.

Application

Fluid A is used to rinse aqueous solutions during sterility testing by membrane filtration. It is also used to dilute soluble solids for the same application. This product is also recommended as a rinsing fluid for membrane filtration of non-sterile products. It is excellent for dissolving or diluting samples, reconstituting commercial microorganisms, or as a transport medium for microorganisms.
Sterility testing
Bioburden testing

Packaging

Fluid A is available as:
STBMRFA12: Pack of 12 bottles, each bottle with 100 mL Fluid A.
STBMRFA34: Pack of 4 bottles, each bottle with 300 mL Fluid A.
STBMRFA64: Pack of 4 bottles, each bottle with 600 mL Fluid A

Components

Screw cap with septum

Culture Medium

Fluid A

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service