MilliporeSigma
All Photos(1)

SX0320

Supelco

Sodium bicarbonate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Sign Into View Organizational & Contract Pricing

Select a Size

Synonym(s):
Sodium Hydrogen Carbonate, Baking Soda
Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
Beilstein:
4153970
MDL number:
EC Index Number:
205-633-8

Agency

meets USP/NF

Quality Level

Assay

99.7-100.3% dry basis

form

powder

impurities

≤0.015% Insoluble matter

pH

8.6 (20 °C, 50 g/L in H2O)

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

mp

270 °C (decomposition)

solubility

96 g/L

density

2.16 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.003%
phosphate (PO43-): ≤0.001%
sulfur compounds (as SO42-): ≤0.003%

cation traces

Ca: ≤0.02%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.005%
NH4+: ≤5 ppm
heavy metals (as Pb): ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
1343371631S8875
Sodium bicarbonate Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Supelco

SX0320

Sodium bicarbonate

-
Sodium bicarbonate puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder

Sigma-Aldrich

13433

Sodium bicarbonate

Essential Grade
Sodium bicarbonate tested according to Ph. Eur.

Sigma-Aldrich

71631

Sodium bicarbonate

-
Sodium bicarbonate ReagentPlus®, ≥99.5%, powder

Sigma-Aldrich

S8875

Sodium bicarbonate

-
form

powder

form

powder

form

solid

form

powder

impurities

≤0.015% Insoluble matter

impurities

ammonia (NH3), in accordance, organic volatile impurities, complies, residual solvents, complies, water-insoluble matter, complies, ≤0.002% ammonium (NH4), complies

impurities

-

impurities

-

pH

8.6 (20 °C, 50 g/L in H2O)

pH

≤8.6 (20 °C, 5%)

pH

-

pH

-

mp

270 °C (decomposition)

mp

-

mp

-

mp

-

solubility

96 g/L

solubility

-

solubility

water: soluble 50 g/L

solubility

-

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Ammonium (NH4): 5ppm max
Assay (dried basis): 99.7-100.3%
Calcium (Ca): 0.02% max
Chloride (Cl): 0.003% max
Heavy metals (as Pb): 5ppm max
Insoluble matter: 0.015% max
Iron (Fe): 0.001% max
Magnesium (Mg): 0.005% max
Phosphate (PO4): 0.001% max
Potassium (K): 0.005% max
Sulfur compounds (as SO4): 0.003% max

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 8

1 of 8

Sodium bicarbonate BioXtra, 99.5-100.5%

Sigma-Aldrich

S6297

Sodium bicarbonate

Sodium bicarbonate tested according to Ph. Eur.

Sigma-Aldrich

71631

Sodium bicarbonate

Sodium bicarbonate anhydrous, free-flowing, Redi-Dri™, ACS reagent, ≥99.7%

Sigma-Aldrich

792519

Sodium bicarbonate

Sodium bicarbonate anhydrous, free-flowing, Redi-Dri™, ReagentPlus®, ≥99.5%

Sigma-Aldrich

900506

Sodium bicarbonate

Sodium chloride Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Supelco

SX0420

Sodium chloride

Magnesium sulfate anhydrous powder Meets Reagent Specifications for testing USP/NF monographs GR

Supelco

MX0075

Magnesium sulfate

Alexandra M Kozlov et al.
Scientific reports, 10(1), 8388-8388 (2020-05-21)
Recent evidence has emerged that cancer cells can use various metabolites as fuel sources. Restricting cultured cancer cells to sole metabolite fuel sources can promote metabolic changes leading to enhanced glycolysis or mitochondrial OXPHOS. However, the effect of metabolite-restriction on non-transformed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service