Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX0694

Supelco

Sodium Perchlorate Monohydrate

Meets ACS Specifications GR ACS

Synonym(s):

Sodium Perchlorate Monohydrate, Perchloric Acid, Sodium Salt, Monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClNaO4 · xH2O
CAS Number:
Molecular Weight:
122.44 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-511-9
assay:
85.0-90.0% (NaClO4 basis)
grade:
ACS reagent
form:
crystals
solubility:
2090 g/L

grade

ACS reagent

Quality Level

assay

85.0-90.0% (NaClO4 basis)

form

crystals

impurities

≤0.005% Insoluble matter

color

white

pH

4.5-7.0 (25 °C, 50 g/L in H2O)
6.0-8.0 (25 °C, 5% in solution)

mp

130 °C

solubility

2090 g/L

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.02%
Fe: ≤5 ppm
K: ≤0.05%
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

InChI

1S/ClHO4.Na.H2O/c2-1(3,4)5;;/h(H,2,3,4,5);;1H2/q;+1;/p-1

InChI key

IXGNPUSUVRTQGW-UHFFFAOYSA-M

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay (as NaClO4): 85.0-90.0%
Calcium (Ca): 0.02% max
Chloride (Cl): 0.003% max
Color: White
Form: Moist crystals
Heavy Metals (by ICP-OES): 5ppm max
Insoluble matter: 0.005% max
Iron (Fe): 5ppm max
Potassium (K): 0.05% max
Sulfate (SO4): 0.002% max
pH of a 5% solution at 25°C: 6.0-8.0

pictograms

Flame over circleExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service