Documents
SYMBA4RP1
Millipore
Steritest® Symbio Essential Requalification Protocol
A4 format
UNSPSC Code:
41116500
NACRES:
NB.79
Quality Level
compatibility
for use with Steritest® Symbio pump.
General description
Steritest® Symbio Essential Requalification protocol is designed as a ready to use guide to perform the requalification of the Steritest® Symbio pump. The requalification protocol can be used to re-certify consistent performance of the pumps upon encountering any change or at regular intervals.
Application
Steritest® Symbio Essential Requalification protocol is used for the re-qualification of Steritest® Symbio pump which is used to perform sterility testing for pharmaceutical products.
Legal Information
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany