Skip to Content
MilliporeSigma

SYMBDSH03

Steritest® Symbio Drain Tray Support

For 3 canisters, LFH pump

Synonym(s):

Canister holder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
23151818
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

316L stainless steel (Drain Tray Support), EPDM rubber seal

agency

complies with EP (2.6.1), complies with JP (4.06), complies with USP (71)

manufacturer/tradename

Steritest®

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio Pump

shipped in

ambient

General description

Steritest® Symbio drain tray support includes the stainless steal canister holder and quick connector designed for 3 canisters and the usage of LFH pump.

Application

Accessories for Streamlining Your Workflow and Increasing Safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library