MilliporeSigma
All Photos(3)

SYMBDTH03

Millipore

Steritest® Symbio Complete Drain Tray

For 3 canisters, LFH pump

Synonym(s):
Outlet

material

316L stainless steel (Drain Tray Support)
EPDM rubber seal
polyphenylsulfone (PPSU) container

Agency

complies with EP (2.6.1)
complies with JP (4.06)
complies with USP (71)

manufacturer/tradename

Steritest®

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio Pump

shipped in

ambient

Compare Similar Items

View Full Comparison

1 of 4

This Item
SYMBDTH01SYMBDTL03SYMBDTL01
material

316L stainless steel (Drain Tray Support), EPDM rubber seal, polyphenylsulfone (PPSU) container

material

316 stainless steel support (drain tray), EPDM rubber seal, polyphenylsulfone (PPSU) container (drain tray)

material

316L stainless steel (Drain Tray Support), EPDM rubber seal

material

316L stainless steel support (drain tray), EPDM rubber seal, polyphenylsulfone (PPSU) container (drain tray)

application(s)

pharmaceutical
sterility testing

application(s)

pharmaceutical
sterility testing

application(s)

pharmaceutical
sterility testing

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio Pump

compatibility

for use with Steritest® Symbio pump

compatibility

for use with Steritest® Symbio Pump

compatibility

for use with Steritest® Symbio pump

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

agency

complies with EP (2.6.1)

agency

EP (2.6.1), JP (4.06), USP (71)

agency

complies with EP (2.6.1), complies with JP (4.06), complies with USP (71)

agency

EP (2.6.1), JP (4.06), USP (71)

manufacturer/tradename

Steritest®

manufacturer/tradename

Steritest®

manufacturer/tradename

Steritest®

manufacturer/tradename

Steritest®

General description

Steritest® Symbio complete drain tray includes stainless steel canister holder, plastic fluid collector, and seal. This tray is designed for 3 canisters and the usage of LFH Pump. The Steritest® Symbio LFH Pump is designed for different testing environments, including a laminar flow hood, biosafety cabinet, cleanroom, or even inside an isolator.

Application

Steritest® Symbio complete drain tray is a Steritest® Symbio LFH Pump accessory for streamlining your workflow and increasing safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service