Key Documents
SYMBLFH01WW
Millipore
Steritest Symbio LFH Pump Kit, 2 media
About This Item
UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
JA.12
This product is sold exclusively by Millipore via offline channels. Contact Customer Service
material
316L stainless steel
Quality Level
product line
Steritest®
weight
14.2 kg (pump body) with standard accessories:15.8 kg
General description
With its compact design, the Steritest Symbio LFH Pump can be used comfortably in the smallest testing environments, including in the laminar flow hood, biosafety cabinet, clean room or even inside an isolator.
Application
The Steritest Symbio LFH Pump, designed to fit your testing environment, Sterility Testing.
Legal Information
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Sterility testing USP 71, EP2.6.1 , JP 4.06