MilliporeSigma
All Photos(1)

TRLABEL01

Millipore

Traceabilty Labels

Sparepart belonging to the Milliflex® Quantum rapid detection system

Sign Into View Organizational & Contract Pricing

Select a Size

Synonym(s):
Barcode label
eCl@ss:
32160606
NACRES:
NB.35

sterility

non-sterile

manufacturer/tradename

Milliflex®

input

pharmaceutical(s)

application(s)

industrial qc
pharmaceutical

shipped in

dry ice

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
MXQLAB001Z743668Z743679
Millipore

Millipore

TRLABEL01

Traceabilty Labels

sterility

non-sterile

sterility

-

sterility

-

sterility

-

input

pharmaceutical(s)

input

-

input

-

input

-

application(s)

industrial qc
pharmaceutical

application(s)

pharmaceutical

application(s)

-

application(s)

-

shipped in

dry ice

shipped in

ambient

shipped in

-

shipped in

-

General description

Traceability labels enable scientists to efficiently keep track of different samples being incubated. Information pertaining to a particular experiment can be recorded as such on the label for easy tracking. Used as part of the Milliflex® Quantum rapid detection system it can used to identify different samples after membrane filtration.

Application

Traceability labels is used to keep track of different samples post membrane filtration when the filtered samples, placed on media plates are left for incubation.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service