Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TX0075

Sigma-Aldrich

α-Terpineol

Synonym(s):

α-Terpineol, Terpineol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3CC6H8COH(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-680-6

vapor pressure

0.056 hPa ( 24 °C)

assay

≥97% (GC)

form

liquid

bp

217-218 °C/1013 hPa

mp

32-34 °C

transition temp

flash point 89 °C

solubility

0.71 g/L

shipped in

ambient

storage temp.

room temp

InChI

1S/C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11/h4,9,11H,5-7H2,1-3H3/t9-/m0/s1

InChI key

WUOACPNHFRMFPN-VIFPVBQESA-N

Analysis Note

Assay (GC): 97% min
Color (APHA): 10 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

195.8 °F - closed cup

flash_point_c

91 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service