Key Documents
ZLXEV100WW
Milli-Q
Elix Essential 10 UV Water Purification System
About This Item
UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
SB.53
product line
Elix® Essential
Quality Level
General description
Elix Essential systems are ideal for laboratories needing an easy-to-use economical solution that provides a constant and reliable source of Type 2 pure water. Patented Elix electrodeionization technology is combined with complementary water purification techniques, including pretreatment, reverse osmosis and bacteriocidal UV lamp treatment (in Elix Essential UV systems) to produce Type 2 pure water of consistently-pure and reliable water quality. As there are no resin packs to change, thanks to the Elix module technology, users benefit from low and predictable running costs.
Application
A pure water solution, integrating Elix technology for your essential needs
Legal Information
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Directions for Use