MilliporeSigma
All Photos(4)

ZRDS0P3WW

Milli-Q

RiOs-DI® Water Purification System

A basic solution for Type 2 pure water production with integrated 6 L storage tank

Sign Into View Organizational & Contract Pricing
Synonym(s):
RiOs-DI® 3 Water Purification System

AC/DC input

100 - 230 V AC, 50 - 60 Hz (per your country requirements)

Quality Level

packaging

pack of 1 unit

parameter

16.7 kg operating weight (36.7 lb)

resistivity

>10 MΩ-cm, at 25°C

H × W × D

49.7 cm (19.6 in.) × 29 cm (11.4 in.) × 33.9 cm (13.3 in.)

tank volume

6 L , Integrated storage tank

input

feed water nature potable tap water

output

product water quality: type 2 water (> 5 MΩ·cm)

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ZRDSVP3WWZR0S0P8WWZR0E0160WW
parameter

16.7 kg operating weight (36.7 lb)

parameter

17.3 kg operating weight (38.0 lb), 3 L/hr flow rate, 40 L/day max. usage (water)

parameter

160 L/day max. usage (water), 8 L/hr flow rate, 8.9 kg operating weight (19.6 lb)

parameter

15.7 kg operating weight (34.5 lb), 16 L/hr flow rate, 300 L/day max. usage (water)

resistivity

>10 MΩ-cm, at 25°C

resistivity

>10 MΩ-cm, at 25°C

resistivity

-

resistivity

-

H × W × D

49.7 cm (19.6 in.) × 29 cm (11.4 in.) × 33.9 cm (13.3 in.)

H × W × D

49.7 cm (19.6 in.) × 29 cm (11.4 in.) × 33.9 cm (13.3 in.)

H × W × D

49.7 cm (19.6 in.) × 29 cm (11.4 in.) × 33.0 cm (13.0 in.)

H × W × D

47 cm (18.5 in.) × 26.8 cm (10.6 in.) × 33.9 cm (13.3 in.)

input

feed water nature potable tap water

input

feed water nature potable tap water

input

feed water nature potable tap water

input

feed water nature potable tap water

output

product water quality: type 2 water (> 5 MΩ·cm)

output

product water quality: type 2 water (> 5 MΩ·cm)

output

product water quality: type 3 water (RO water)

output

product water quality: type 3 water (RO water)

General description

With a combination of reverse osmosis (RO) technology and deionization (DI) resins, the compact, user-friendly RiOs-DI® 3 Water Purification System produces up to 15 L per day of consistent-quality Type 2 pure water—easily and economically.
The combined power of RO and DI resins provides pure water with high resistivity (> 10 MΩ·cm) and low TOC (< 30 ppb) whose quality surpasses that of service DI or distilled water. RiOs-DI® systems let you avoid the many drawbacks of these other purification methods: high lab space demands to store resin bottles or accommodate distillation equipment; heavy maintenance; lack of water quality monitoring; and in the case of distillation, high energy and water use, as well as the need for strong chemicals for cleaning.

Application

RiOs-DI® systems are a source of pure (Type 2) water for general laboratory tasks, such as preparation of buffers, microbiological culture media and reagent solutions, and rinsing of laboratory glassware.

Features and Benefits

  • Superior quality pure water: Easy-to-use RiOs-DI® systems produce Type 2 pure water with quality superior to that of service DI or stills. With their combination of reverse osmosis (RO) technology and deionization resins, RiOs-DI® systems let you avoid the issues associated with service DI or distillation (maintenance, storage, cleaning, etc.).
  • Pure water for general laboratory use: Water from RiOs-DI® systems is suitable for a variety of uses, including buffer and reagent preparation; microbiological culture media preparation; and glassware rinsing.
  • Compact design for the most efficient use of your lab space: A small footprint makes it easy to install the RiOs-DI® system wherever you want to in your lab.
  • Flow rate and storage volume to meet your low-volume pure water needs: Systems have a pure water flow rate of >2.4 L per hour and an integrated 6 L reservoir for pure water storage.
  • High-quality water for applications requiring low bacterial levels: An optional built-in bactericidal UV lamp is available to help ensure optimal water quality for bacteria-sensitive applications.
  • Easily accessible information on system operation: The user-friendly display provides system status at a glance; the concise Quick Reference Guide is a handy guide for daily operation.
  • Easy installation: The system is designed for easy installation. Just connect the system to a tap water supply, plug it in, and insert the SmartPak® RODI cartridge -your system is ready to use!
  • Simple self-maintenance: The single SmartPak® RODI cartridge enables easy and rapid replacement.

Components

  • This is a web reference. The end letters "WW" will be replaced by the country letters on your invoice to have the right shipping kit for the country. This ensures you receive the user manual in local language and a power cord adapted to the local electrical network.
  • The indicated price currently corresponds to the above catalogue number only; it is subject to change at any time without notice.
  • This catalogue number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you; do not hesitate to contact us!

Other Notes

For more information on this product please contact our Technical Service Team

Legal Information

RIOS-DI is a registered trademark of Merck KGaA, Darmstadt, Germany
SMARTPAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service